4'-Chlor-3-(1,3-dioxolan-2-yl)propiophenon

Lieferant: FLUOROCHEM

022289-1G
FLUO022289-1GEA 0 EUR
FLUO022289-1G
4'-Chlor-3-(1,3-dioxolan-2-yl)propiophenon
4'-Chlor-3-(1,3-dioxolan-2-yl)propiophenon
Formel: C₁₂H₁₃ClO₃
MDL: MFCD06201165
CAS-Nummer: 147030-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR