3'-Chlor-3-(1,3-dioxolan-2-yl)propiophenon

Lieferant: FLUOROCHEM

022290-1G
FLUO022290-1GEA 0 EUR
FLUO022290-1G
3'-Chlor-3-(1,3-dioxolan-2-yl)propiophenon
3'-Chlor-3-(1,3-dioxolan-2-yl)propiophenon
Formel: C₁₂H₁₃ClO₃
MDL: MFCD06201166
CAS-Nummer: 147030-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR