3-(1,3-Dioxolan-2-yl)-4'-fluorpropiophenon

Lieferant: FLUOROCHEM

022294-1G
FLUO022294-1GEA 0 EUR
FLUO022294-1G
3-(1,3-Dioxolan-2-yl)-4'-fluorpropiophenon
3-(1,3-Dioxolan-2-yl)-4'-fluorpropiophenon
Formel: C₁₂H₁₃FO₃
MDL: MFCD06201169
CAS-Nummer: 116332-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR