3-(1,3-Dioxolan-2-yl)-3'-fluorpropiophenon

Lieferant: FLUOROCHEM

022295-1G
FLUO022295-1GEA 0 EUR
FLUO022295-1G
3-(1,3-Dioxolan-2-yl)-3'-fluorpropiophenon
3-(1,3-Dioxolan-2-yl)-3'-fluorpropiophenon
Formel: C₁₂H₁₃FO₃
MDL: MFCD06201170
CAS-Nummer: 842124-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR