3-(1,3-Dioxolan-2-yl)-4'-propylpropiophenon

Lieferant: FLUOROCHEM

022300-1G
FLUO022300-1GEA 0 EUR
FLUO022300-1G
3-(1,3-Dioxolan-2-yl)-4'-propylpropiophenon
3-(1,3-Dioxolan-2-yl)-4'-propylpropiophenon
Formel: C₁₅H₂₀O₃
MDL: MFCD06201175
CAS-Nummer: 842124-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR