3-(1,3-Dioxolan-2-yl)-4'-methoxypropiophenon

Lieferant: FLUOROCHEM

022304-1G
FLUO022304-1GEA 0 EUR
FLUO022304-1G
3-(1,3-Dioxolan-2-yl)-4'-methoxypropiophenon
3-(1,3-Dioxolan-2-yl)-4'-methoxypropiophenon
Formel: C₁₃H₁₆O₄
MDL: MFCD06201177
CAS-Nummer: 147030-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR