3-(1,3-Dioxolan-2-yl)-3'-methoxypropiophenon
Lieferant: FLUOROCHEM
022305-1G
FLUO022305-1GEA
0
EUR
FLUO022305-1G
3-(1,3-Dioxolan-2-yl)-3'-methoxypropiophenon
3-(1,3-Dioxolan-2-yl)-3'-methoxypropiophenon
Formel:
C₁₃H₁₆O₄ |
MDL:
MFCD06201178 CAS-Nummer: 842124-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...