3,5-Dimethylbenzaldehyd

Lieferant: FLUOROCHEM

022364-1G
FLUO022364-1GEA 0 EUR
FLUO022364-1G
3,5-Dimethylbenzaldehyd
3,5-Dimethylbenzaldehyde
Formel: C₉H₁₀O
Molecular Weight: 134,18 g/mol
Dichte: 1,09 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00082777
CAS-Nummer: 5779-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR