4-(4-Chlor-3-fluorphenyl)benzaldehyd

Lieferant: FLUOROCHEM

Synonyme: 4'-Chloro-3'-fluoro-[1,1'-biphenyl]-4-carboxaldehyde, 4'-Chloro-3'-fluoro[1,1'-biphenyl]-4-carbaldehyde

022365-1G
FLUO022365-1GEA 0 EUR
FLUO022365-1G
4-(4-Chlor-3-fluorphenyl)benzaldehyd
4-(4-Chloro-3-fluorophenyl)benzaldehyde
Formel: C₁₃H₈ClFO
MDL: MFCD06201223
CAS-Nummer: 842140-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR