3-(4-Chlor-3-fluorphenyl)benzaldehyd

Lieferant: FLUOROCHEM

022366-1G
FLUO022366-1GEA 0 EUR
FLUO022366-1G
3-(4-Chlor-3-fluorphenyl)benzaldehyd
3-(4-Chloro-3-fluorophenyl)benzaldehyde
Formel: C₁₃H₈ClFO
Molecular Weight: 234.65 g/mol
MDL: MFCD06201224
CAS-Nummer: 842140-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR