5-Fluor-2-methylbenzaldehyd

Lieferant: FLUOROCHEM

022378-1G
FLUO022378-1GEA 31.1 EUR
FLUO022378-1G
5-Fluor-2-methylbenzaldehyd
5-Fluor-2-methylbenzaldehyd
Formel: C₈H₇FO
Molecular Weight: 138,14 g/mol
MDL: MFCD02261763
CAS-Nummer: 22062-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR