3,4-Difluorbenzoylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 3-(3,4-Difluorophenyl)-3-oxopropanenitrile

022381-1G
FLUO022381-1GEA 228 EUR
FLUO022381-1G
3,4-Difluorbenzoylacetonitril
3,4-Difluorbenzoylacetonitril
Formel: C₉H₅F₂NO
Molecular Weight: 181,14 g/mol
CAS-Nummer: 71682-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR