3-Fluorbenzoylacetonitril
Lieferant: FLUOROCHEM
Synonyme:
3-(3-Fluorophenyl)-3-oxopropanenitrile
022382-1G
FLUO022382-1GEA
143
EUR
FLUO022382-1G
3-Fluorbenzoylacetonitril
3-Fluorbenzoylacetonitril
Formel:
C₉H₆FNO Molecular Weight: 163,15 g/mol |
MDL:
MFCD02260777 CAS-Nummer: 21667-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...