3,5-Difluorbenzoylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 3-(3,5-Difluorophenyl)-3-oxopropanenitrile

022383-1G
FLUO022383-1GEA 594 EUR
FLUO022383-1G
3,5-Difluorbenzoylacetonitril
3,5-Difluorbenzoylacetonitril
Formel: C₉H₅F₂NO
Molecular Weight: 181,14 g/mol
MDL: MFCD02260784
CAS-Nummer: 842140-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR