3,5-Dichlorbenzoylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 3-(3,5-Dichlorophenyl)-3-oxopropanenitrile

022385-1G
FLUO022385-1GEA 209 EUR
FLUO022385-1G
3,5-Dichlorbenzoylacetonitril
3,5-Dichlorbenzoylacetonitril
Formel: C₉H₅Cl₂NO
CAS-Nummer: 69316-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR