4-Fluorbenzoylacetonitril

Lieferant: FLUOROCHEM

Synonyme: 3-(4-Fluorphenyl)-3-oxopropionsäurenitril, 3-(4-Fluorphenyl)-3-ketopropionsäurenitril

022389-1G
FLUO022389-1GEA 264 EUR
FLUO022389-1G
4-Fluorbenzoylacetonitril
4-Fluorbenzoylacetonitril
Formel: C₉H₆FNO
Molecular Weight: 163,15 g/mol
MDL: MFCD00662062
CAS-Nummer: 4640-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR