4'-Propylacetophenon
Lieferant: FLUOROCHEM
Synonyme:
4-Propyl-1-acetylbenzol, 1-(4-Propylphenyl)ethanon, p-Propylacetophenon
022390-1G
FLUO022390-1GEA
60
EUR
FLUO022390-1G
4'-Propylacetophenon
4'-Propylacetophenon
Formel:
C₁₁H₁₄O Molecular Weight: 162,23 g/mol |
MDL:
MFCD00041359 CAS-Nummer: 2932-65-2 EINECS: 220-903-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...