3',5'-Dichloracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(3,5-Dichlorophenyl)ethanone
022391-1G
FLUO022391-1GEA
34.1
EUR
FLUO022391-1G
3',5'-Dichloracetophenon
3',5'-Dichloracetophenon
Formel:
C₈H₆Cl₂O Molecular Weight: 189.04 g/mol |
MDL:
MFCD00045189 CAS-Nummer: 14401-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...