3'-Chlor-5'-fluoracetophenon
Lieferant: FLUOROCHEM
022392-1G
FLUO022392-1GEA
137
EUR
FLUO022392-1G
3'-Chlor-5'-fluoracetophenon
3'-Chlor-5'-fluoracetophenon
Formel:
C₈H₆ClFO |
MDL:
MFCD04115834 CAS-Nummer: 842140-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...