4'-(Ethylthio)acetophenon
Lieferant: FLUOROCHEM
022394-1G
FLUO022394-1GEA
104
EUR
FLUO022394-1G
4'-(Ethylthio)acetophenon
4'-(Ethylthio)acetophenon
Formel:
C₁₀H₁₂OS Molecular Weight: 180,27 g/mol |
MDL:
MFCD06201229 CAS-Nummer: 3585-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...