4'-Isopropylacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Isopropylphenyl)ethanon, 4-Isopropyl-1-acetylbenzol, p-Isopropylacetophenon

022395-1G
FLUO022395-1GEA 0 EUR
FLUO022395-1G
4'-Isopropylacetophenon
4'-Isopropylacetophenon
Formel: C₁₁H₁₄O
Molecular Weight: 162,23 g/mol
Dichte: 0,97 g/cm³ (20 °C)
MDL: MFCD00048297
CAS-Nummer: 645-13-6
EINECS: 211-433-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR