3,4'-Dimethylbenzhydrol
Lieferant: FLUOROCHEM
Synonyme:
3-Methyl-α-(4-methylphenyl)benzenemethanol
022396-1G
FLUO022396-1GEA
1050
EUR
FLUO022396-1G
3,4'-Dimethylbenzhydrol
3,4'-Dimethylbenzhydrol
Formel:
C₁₅H₁₆O Molecular Weight: 212,29 g/mol |
MDL:
MFCD06201230 CAS-Nummer: 13389-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...