3-Chlor-4-fluor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022398-1G
FLUO022398-1GEA 1050 EUR
FLUO022398-1G
3-Chlor-4-fluor-4'-methylbenzhydrol
3-Chlor-4-fluor-4'-methylbenzhydrol
Formel: C₁₄H₁₂ClFO
MDL: MFCD06201232
CAS-Nummer: 842140-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR