4-Chlor-3-fluor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022400-1G
FLUO022400-1GEA 1050 EUR
FLUO022400-1G
4-Chlor-3-fluor-4'-methylbenzhydrol
4-Chlor-3-fluor-4'-methylbenzhydrol
Formel: C₁₄H₁₂ClFO
MDL: MFCD06201234
CAS-Nummer: 842140-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR