3,4-Dichlor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022402-1G
FLUO022402-1GEA 1050 EUR
FLUO022402-1G
3,4-Dichlor-4'-methylbenzhydrol
3,4-Dichlor-4'-methylbenzhydrol
Formel: C₁₄H₁₂Cl₂O
MDL: MFCD06201236
CAS-Nummer: 842140-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR