3,5-Dichlor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022403-1G
FLUO022403-1GEA 1050 EUR
FLUO022403-1G
3,5-Dichlor-4'-methylbenzhydrol
3,5-Dichlor-4'-methylbenzhydrol
Formel: C₁₄H₁₂Cl₂O
MDL: MFCD06201237
CAS-Nummer: 13395-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR