3,4-Difluor-4'-methylbenzhydrol
Lieferant: FLUOROCHEM
022404-1G
FLUO022404-1GEA
1050
EUR
FLUO022404-1G
3,4-Difluor-4'-methylbenzhydrol
3,4-Difluor-4'-methylbenzhydrol
Formel:
C₁₄H₁₂F₂O |
MDL:
MFCD06201238 CAS-Nummer: 842140-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...