3,5-Difluor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022405-1G
FLUO022405-1GEA 1050 EUR
FLUO022405-1G
3,5-Difluor-4'-methylbenzhydrol
3,5-Difluor-4'-methylbenzhydrol
Formel: C₁₄H₁₂F₂O
MDL: MFCD06201239
CAS-Nummer: 842140-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR