3,4-Dimethoxy-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022406-1G
FLUO022406-1GEA 1040 EUR
FLUO022406-1G
3,4-Dimethoxy-4'-methylbenzhydrol
3,4-Dimethoxy-4'-methylbenzhydrol
Formel: C₁₆H₁₈O₃
CAS-Nummer: 423177-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR