4-(Dimethylamino)-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022407-1G
FLUO022407-1GEA 1040 EUR
FLUO022407-1G
4-(Dimethylamino)-4'-methylbenzhydrol
4-(Dimethylamino)-4'-methylbenzhydrol
Formel: C₁₆H₁₉NO
CAS-Nummer: 93026-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR