3-Fluor-4'-methylbenzhydrol

Lieferant: FLUOROCHEM

022411-1G
FLUO022411-1GEA 1010 EUR
FLUO022411-1G
3-Fluor-4'-methylbenzhydrol
3-Fluor-4'-methylbenzhydrol
Formel: C₁₄H₁₃FO
CAS-Nummer: 38158-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR