3-Methoxy-4'-methylbenzhydrol
Lieferant: FLUOROCHEM
Synonyme:
3-Methoxy-α-(4-methylphenyl)benzenemethanol
022412-1G
FLUO022412-1GEA
1010
EUR
FLUO022412-1G
3-Methoxy-4'-methylbenzhydrol
3-Methoxy-4'-methylbenzhydrol
Formel:
C₁₅H₁₆O₂ Molecular Weight: 228,29 g/mol |
MDL:
MFCD06201245 CAS-Nummer: 842140-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...