4-Methoxy-4'-methylbenzhydrol
Lieferant: FLUOROCHEM
022413-1G
FLUO022413-1GEA
1010
EUR
FLUO022413-1G
4-Methoxy-4'-methylbenzhydrol
4-Methoxy-4'-methylbenzhydrol
Formel:
C₁₅H₁₆O₂ |
CAS-Nummer:
383-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...