4-Methyl-4'-(methylthio)benzhydrol

Lieferant: FLUOROCHEM

022414-1G
FLUO022414-1GEA 1010 EUR
FLUO022414-1G
4-Methyl-4'-(methylthio)benzhydrol
4-Methyl-4'-(methylthio)benzhydrol
Formel: C₁₅H₁₆OS
CAS-Nummer: 842140-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR