4-tert-Butylbenzhydrol

Lieferant: FLUOROCHEM

Synonyme: (4-(tert-Butyl)phenyl)(phenyl)methanol

022415-1G
FLUO022415-1GEA 279 EUR
FLUO022415-1G
4-tert-Butylbenzhydrol
4-tert-Butylbenzhydrol
Formel: C₁₇H₂₀O
Molecular Weight: 240,34 g/mol
MDL: MFCD06201248
CAS-Nummer: 22543-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR