3-Chlor-4-fluorbenzhydrol

Lieferant: FLUOROCHEM

022416-1G
FLUO022416-1GEA 528 EUR
FLUO022416-1G
3-Chlor-4-fluorbenzhydrol
3-Chlor-4-fluorbenzhydrol
Formel: C₁₃H₁₀ClFO
MDL: MFCD06201249
CAS-Nummer: 842140-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR