3-Chlor-5-fluorbenzhydrol

Lieferant: FLUOROCHEM

022417-1G
FLUO022417-1GEA 1010 EUR
FLUO022417-1G
3-Chlor-5-fluorbenzhydrol
3-Chlor-5-fluorbenzhydrol
Formel: C₁₃H₁₀ClFO
Molecular Weight: 236,669 g/mol
MDL: MFCD06201250
CAS-Nummer: 842140-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR