4-Chlor-3-fluorbenzhydrol
Lieferant: FLUOROCHEM
022418-1G
FLUO022418-1GEA
1010
EUR
FLUO022418-1G
4-Chlor-3-fluorbenzhydrol
4-Chlor-3-fluorbenzhydrol
Formel:
C₁₃H₁₀ClFO Molecular Weight: 236,669 g/mol |
MDL:
MFCD06201251 CAS-Nummer: 842140-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...