3,5-Dichlorbenzhydrol

Lieferant: FLUOROCHEM

022421-1G
FLUO022421-1GEA 531 EUR
FLUO022421-1G
3,5-Dichlorbenzhydrol
3,5-Dichlorbenzhydrol
Formel: C₁₃H₉Cl₃O
MDL: MFCD06201254
CAS-Nummer: 13395-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR