3,4-Difluorbenzhydrol

Lieferant: FLUOROCHEM

022422-1G
FLUO022422-1GEA 1010 EUR
FLUO022422-1G
3,4-Difluorbenzhydrol
3,4-Difluorbenzhydrol
Formel: C₁₃H₁₀F₂O
MDL: MFCD06201255
CAS-Nummer: 182192-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR