3,5-Difluorbenzhydrol
Lieferant: FLUOROCHEM
022423-1G
FLUO022423-1GEA
1010
EUR
FLUO022423-1G
3,5-Difluorbenzhydrol
3,5-Difluorbenzhydrol
Formel:
C₁₃H₁₀F₂O Molecular Weight: 220,22 g/mol |
MDL:
MFCD06201256 CAS-Nummer: 153877-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...