3,4-Dimethoxybenzhydrol
Lieferant: FLUOROCHEM
022424-1G
FLUO022424-1GEA
656
EUR
FLUO022424-1G
3,4-Dimethoxybenzhydrol
3,4-Dimethoxybenzhydrol
Formel:
C₁₅H₁₆O₃ |
CAS-Nummer:
56139-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...