4-(Dimethylamino)benzhydrol
Lieferant: FLUOROCHEM
Synonyme:
(4-(dimethylamino)phenyl)(phenyl)methanol
022425-1G
FLUO022425-1GEA
1010
EUR
FLUO022425-1G
4-(Dimethylamino)benzhydrol
4-(Dimethylamino)benzhydrol
Formel:
C₁₅H₁₇NO Molecular Weight: 227,302 g/mol |
MDL:
MFCD00963078 CAS-Nummer: 7494-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...