3,5-Dimethylbenzhydrol

Lieferant: FLUOROCHEM

022426-1G
FLUO022426-1GEA 1010 EUR
FLUO022426-1G
3,5-Dimethylbenzhydrol
3,5-Dimethylbenzhydrol
Formel: C₁₆H₁₈O
CAS-Nummer: 13389-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR