4-(Ethylthio)benzhydrol
Lieferant: FLUOROCHEM
022428-1G
FLUO022428-1GEA
1200
EUR
FLUO022428-1G
4-(Ethylthio)benzhydrol
4-(Ethylthio)benzhydrol
Formel:
C₁₅H₁₆OS |
MDL:
MFCD06201259 CAS-Nummer: 842140-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...