3-Fluorbenzhydrol
Lieferant: FLUOROCHEM
Synonyme:
(3-Fluorophenyl)(phenyl)methanol
022429-1G
FLUO022429-1GEA
284
EUR
FLUO022429-1G
3-Fluorbenzhydrol
3-Fluorbenzhydrol
Formel:
C₁₃H₁₁FO |
CAS-Nummer:
365-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...