4-(Methylthio)benzhydrol

Lieferant: FLUOROCHEM

022431-1G
FLUO022431-1GEA 1010 EUR
FLUO022431-1G
4-(Methylthio)benzhydrol
4-(Methylthio)benzhydrol
Formel: C₁₄H₁₄OS
MDL: MFCD00093026
CAS-Nummer: 183729-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR