3,3'-Dimethylbenzhydrol

Lieferant: FLUOROCHEM

022432-1G
FLUO022432-1GEA 483 EUR
FLUO022432-1G
3,3'-Dimethylbenzhydrol
3,3'-Dimethylbenzhydrol
Formel: C₁₅H₁₆O
MDL: MFCD06201262
CAS-Nummer: 13389-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR