3-Chlor-4-fluor-3'-methylbenzhydrol

Lieferant: FLUOROCHEM

022434-1G
FLUO022434-1GEA 1200 EUR
FLUO022434-1G
3-Chlor-4-fluor-3'-methylbenzhydrol
3-Chlor-4-fluor-3'-methylbenzhydrol
Formel: C₁₄H₁₂ClFO
CAS-Nummer: 842140-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR