3-Chlor-5-fluor-3'-methylbenzhydrol

Lieferant: FLUOROCHEM

022435-1G
FLUO022435-1GEA 1200 EUR
FLUO022435-1G
3-Chlor-5-fluor-3'-methylbenzhydrol
3-Chlor-5-fluor-3'-methylbenzhydrol
Formel: C₁₄H₁₂ClFO
MDL: MFCD06201265
CAS-Nummer: 842140-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR